Learn More
Abnova™ Human HS3ST2 Partial ORF (NP_006034, 268 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009956-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. This gene is expressed predominantly in brain and may play a role in the nervous system. [provided by RefSeq]
Sequence: QIHFVSGERLITDPAGEMGRVQDFLGIKRFITDKHFYFNKTKGFPCLKKTESSLLPRCLGKSKGRTHVQIDPEVIDQLREFYRPYNIKFYETVGQDFRWESpecifications
NP_006034 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QIHFVSGERLITDPAGEMGRVQDFLGIKRFITDKHFYFNKTKGFPCLKKTESSLLPRCLGKSKGRTHVQIDPEVIDQLREFYRPYNIKFYETVGQDFRWE | |
RUO | |
HS3ST2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9956 | |
HS3ST2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
30ST2/3OST2 | |
HS3ST2 | |
Recombinant | |
wheat germ expression system |