missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HS3ST1 Full-length ORF (NP_005105.1, 1 a.a. - 307 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009957-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194272

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It possesses both heparan sulfate glucosaminyl 3-O-sulfotransferase activity, anticoagulant heparan sulfate conversion activity, and is a rate limiting enzyme for synthesis of anticoagulant heparan. This enzyme is an intraluminal Golgi resident protein. [provided by RefSeq]

Sequence: MAALLLGAVLLVAQPQLVPSRPAELGQQELLRKAGTLQDDVRDGVAPNGSAQQLPQTIIIGVRKGGTRALLEMLSLHPDVAAAENEVHFFDWEEHYSHGLGWYLSQMPFSWPHQLTVEKTPAYFTSPKVPERVYSMNPSIRLLLILRDPSERVLSDYTQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLRHIHIVDGDRLIRDPFPEIQKVERFLKLSPQINASNFYFNKTKGFYCLRDSGRDRCLHESKGRAHPQVDPKLLNKLHEYFHEPNKKFFELVGRTFDWH
Specifications

Specifications

NP_005105.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
3OST/3OST1
HS3ST1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9957
HS3ST1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAALLLGAVLLVAQPQLVPSRPAELGQQELLRKAGTLQDDVRDGVAPNGSAQQLPQTIIIGVRKGGTRALLEMLSLHPDVAAAENEVHFFDWEEHYSHGLGWYLSQMPFSWPHQLTVEKTPAYFTSPKVPERVYSMNPSIRLLLILRDPSERVLSDYTQVFYNHMQKHKPYPSIEEFLVRDGRLNVDYKALNRSLYHVHMQNWLRFFPLRHIHIVDGDRLIRDPFPEIQKVERFLKLSPQINASNFYFNKTKGFYCLRDSGRDRCLHESKGRAHPQVDPKLLNKLHEYFHEPNKKFFELVGRTFDWH
RUO
HS3ST1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers