Learn More
Abnova™ Human HPS1 Full-length ORF (NP_872575.1, 1 a.a. - 194 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003257-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is a component of three different protein complexes termed biogenesis of lysosome-related organelles complex (BLOC)-3, BLOC4, and BLOC5. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 1. Multiple transcript variants encoding distinct isoforms have been identified for this gene; the full-length sequences of some of these have not been determined yet. [provided by RefSeq]
Sequence: MKCVLVATEGAEVLFYWTDQEFEESLRLKFGQSENEEEELPALEDQLSTLLAPVIISSMTMLEKLSDTYTCFSTENGNFLYVLHLFGECLFIAINGDHTESEGDLRRKLYVLKYLFEVHFGLVTVDGHLIRKELRPPDLAQRVQLWEHFQSLLWTYSRLREQEQCFAVEVTTGRGSPQDLKLSGPGATDSPPALSpecifications
NP_872575.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HPS/MGC5277 | |
HPS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3257 | |
HPS1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKCVLVATEGAEVLFYWTDQEFEESLRLKFGQSENEEEELPALEDQLSTLLAPVIISSMTMLEKLSDTYTCFSTENGNFLYVLHLFGECLFIAINGDHTESEGDLRRKLYVLKYLFEVHFGLVTVDGHLIRKELRPPDLAQRVQLWEHFQSLLWTYSRLREQEQCFAVEVTTGRGSPQDLKLSGPGATDSPPAL | |
RUO | |
HPS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |