missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HEATR2 Full-length ORF (AAH10850.1, 1 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054919-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MRLKLFSILSTVLLRATDTINSQGQFPSYLETVTKDILAPNLQWHAGRTAAAIRTAAVSCLWALTSSEVLSAEQIRDVQETLMPQVLTTLEEDSKMTRLISCRIINTFLKTSGGMTDPEKLIKIYPELLKRLDDVSNDVRMAAASTLVTWLQCVKGANAKSYYQSSVQYLYRELLVHLDDPERAIQDAILDVPGSPSPSAMIGSFLRPPHPWRTVSQLNLFSSSpecifications
AAH10850.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20397/FLJ25564/FLJ31671/FLJ39381 | |
HEATR2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54919 | |
HEATR2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRLKLFSILSTVLLRATDTINSQGQFPSYLETVTKDILAPNLQWHAGRTAAAIRTAAVSCLWALTSSEVLSAEQIRDVQETLMPQVLTTLEEDSKMTRLISCRIINTFLKTSGGMTDPEKLIKIYPELLKRLDDVSNDVRMAAASTLVTWLQCVKGANAKSYYQSSVQYLYRELLVHLDDPERAIQDAILDVPGSPSPSAMIGSFLRPPHPWRTVSQLNLFSS | |
RUO | |
HEATR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |