missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HDGF Full-length ORF (NP_004485.1, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003068-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118687

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the hepatoma-derived growth factor family. The encoded protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation. This gene was thought initially to be located on chromosome X, however, that location has been determined to correspond to a related pseudogene. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: MSRSNRQKEYKCGDLVFAKMKGYPHWPARIDEMPEAAVKSTANKYQVFFFGTHETAFLGPKDLFPYEESKEKFGKPNKRKGFSEGLWEIENNPTVKASGYQSSQKKSCVEEPEPEPEAAEGDGDKKGNAEGSSDEEGKLVIDEPAKEKNEKGALKRRAGDLLEDSPKRPKEAENPEGEEKEAATLEVERPLPMEVEKNSTPSEPGSGRGPPQEEEEEEDEEEEATKEDAEAPGIRDHESL
Specifications

Specifications

NP_004485.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686J1764/FLJ96580/HMG1L2
HDGF
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
3068
HDGF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSRSNRQKEYKCGDLVFAKMKGYPHWPARIDEMPEAAVKSTANKYQVFFFGTHETAFLGPKDLFPYEESKEKFGKPNKRKGFSEGLWEIENNPTVKASGYQSSQKKSCVEEPEPEPEAAEGDGDKKGNAEGSSDEEGKLVIDEPAKEKNEKGALKRRAGDLLEDSPKRPKEAENPEGEEKEAATLEVERPLPMEVEKNSTPSEPGSGRGPPQEEEEEEDEEEEATKEDAEAPGIRDHESL
RUO
HDGF
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers