missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HCLS1 Partial ORF (AAH16758, 266 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003059-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136041

  • 5420.23 NOK / 25µg

Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: QERKAVTKRSPEAPQPVIAMEEPAVPAPLPKKISSEAWPPVGTPPSSESEPVRTSREHPVPLLPIRQTLPEDNEEPPALPPRTLEGLQVE
Specifications

Specifications

AAH16758
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QERKAVTKRSPEAPQPVIAMEEPAVPAPLPKKISSEAWPPVGTPPSSESEPVRTSREHPVPLLPIRQTLPEDNEEPPALPPRTLEGLQVE
RUO
HCLS1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3059
HCLS1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CTTNL/HS1
HCLS1
Yes
Videos
SDS
Documents
Certificates