Learn More
Abnova™ Human HBS1L Full-length ORF (NP_006611.1, 1 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010767-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq]
Sequence: MARHRNVRGYNYDEDFEDDDLYGQSVEDDYCISPSTAAQFIYSRRDKPSVEPVEEYDYEDLKESSNSVSNHQLSGFDQARLYSCLDHMREVLGDAVPDEILIEAVLKNKFDVQKALSGVLEQDRVQSLKDKNEATVSTGKIAKGKPVDSQTSRSESEIVPKVAKMTVSGKKQTMGFEVPGVSSEENGHSFHTPQKGPPIEDAIASSDVLETASKSANPPHTIQASEEQSSTPAPVKKSGKLRQQIDVKAELEKRQGGKQLLNLVVIGHVDAGKSTLMGHMLYLLGNINKRTMHKYEQESKKAGKASFAYAWVLDETGEERERGVTMDVGMTKFETTTKVITLMDAPGHKDFIPNMITGAAQADVAVLVVDASRGEFEAGFETGGQTREHGLLVRSLGVTQLAVAVNKMDQVNWQQERFQEITGKLGHFLKQAGFKESDVGFIPTSGLSGENLITRSQSSELTKWYKGLCLLEQIDSFKPPQRSIDKPFRLCVSDVFKDQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLHDEPVDWAAAGDHVSLTLVGMDIIKINVGCIFCGPKVPIKACTRFRARILIFNIEIPITKGFPVLLHYQTVSEPAVIKRLISVLNKSTGEVTKKKPKFLTKGQNALVELQTQRPIALELYKDFKELGRFMLRYGGSTIAAGVVTEIKESpecifications
NP_006611.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686L13262/EF-1a/ERFS/HBS1/HSPC276 | |
HBS1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10767 | |
HBS1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MARHRNVRGYNYDEDFEDDDLYGQSVEDDYCISPSTAAQFIYSRRDKPSVEPVEEYDYEDLKESSNSVSNHQLSGFDQARLYSCLDHMREVLGDAVPDEILIEAVLKNKFDVQKALSGVLEQDRVQSLKDKNEATVSTGKIAKGKPVDSQTSRSESEIVPKVAKMTVSGKKQTMGFEVPGVSSEENGHSFHTPQKGPPIEDAIASSDVLETASKSANPPHTIQASEEQSSTPAPVKKSGKLRQQIDVKAELEKRQGGKQLLNLVVIGHVDAGKSTLMGHMLYLLGNINKRTMHKYEQESKKAGKASFAYAWVLDETGEERERGVTMDVGMTKFETTTKVITLMDAPGHKDFIPNMITGAAQADVAVLVVDASRGEFEAGFETGGQTREHGLLVRSLGVTQLAVAVNKMDQVNWQQERFQEITGKLGHFLKQAGFKESDVGFIPTSGLSGENLITRSQSSELTKWYKGLCLLEQIDSFKPPQRSIDKPFRLCVSDVFKDQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLHDEPVDWAAAGDHVSLTLVGMDIIKINVGCIFCGPKVPIKACTRFRARILIFNIEIPITKGFPVLLHYQTVSEPAVIKRLISVLNKSTGEVTKKKPKFLTKGQNALVELQTQRPIALELYKDFKELGRFMLRYGGSTIAAGVVTEIKE | |
RUO | |
HBS1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |