missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GUP1 Partial ORF (NP_065758.2, 157 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057467-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: LISWQSGFVTGTFDLQEVLFHGGSSFTVLRCTSFALESCAHPDRHYSLADSpecifications
NP_065758.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.24kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LISWQSGFVTGTFDLQEVLFHGGSSFTVLRCTSFALESCAHPDRHYSLAD | |
RUO | |
HHATL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57467 | |
GUP1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C3orf3/GUP1/KIAA1173/MBOAT3/MSTP002/OACT3 | |
HHATL | |
Recombinant | |
wheat germ expression system |