Learn More
Abnova™ Human GULP1 Full-length ORF (AAH01103.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00051454-P01.10ug
Weitere Details : Weight : 0.00010kg
Beschreibung
The prompt clearance of cells undergoing apoptosis is critical during embryonic development, normal tissue turnover, inflammation, and autoimmunity. CED6 is an evolutionarily conserved adaptor protein required for efficient engulfment of apoptotic cells by phagocytes.[supplied by OMIM]
Sequence: MNRAFSRKKDKTWMHTPEALSKHFIPYNAKFLGSTEVEQPKGTEVVRDAVRKLKFARHIKKSEGQKIPKVELQISIYGVKILEPKTKEVQHNCQLHRISFCADDKTDKRIFTFICKDSESNKHLCYVFDSEKCVSIPDVVGWFVLFYKPGIVLLLALAKYLKMNNFLSpezifikation
AAH01103.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CED-6/CED6/FLJ31156/GULP | |
GULP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51454 | |
GULP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNRAFSRKKDKTWMHTPEALSKHFIPYNAKFLGSTEVEQPKGTEVVRDAVRKLKFARHIKKSEGQKIPKVELQISIYGVKILEPKTKEVQHNCQLHRISFCADDKTDKRIFTFICKDSESNKHLCYVFDSEKCVSIPDVVGWFVLFYKPGIVLLLALAKYLKMNNFL | |
RUO | |
GULP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |