missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GTPBP3 Partial ORF (NP_116009.1, 244 a.a. - 343 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084705-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: QRLRSGAHVVVTGPPNAGKSSLVNLLSRKPVSIVSPEPGTTRDVLETPVDLAGFPVLLSDTAGLREGVGPVEQEGVRRARERLEQADLILAMLDASDLASSpecifications
NP_116009.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QRLRSGAHVVVTGPPNAGKSSLVNLLSRKPVSIVSPEPGTTRDVLETPVDLAGFPVLLSDTAGLREGVGPVEQEGVRRARERLEQADLILAMLDASDLAS | |
RUO | |
GTPBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84705 | |
GTPBP3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14700/GTPBG3/MSS1/MTGP1/THDF1 | |
GTPBP3 | |
Recombinant | |
wheat germ expression system |