missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GTF3C4 Partial ORF (NP_036336, 723 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009329-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120386

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: DDRTARVLIGHISKKMNKQTFPEHCSLCKEILPFTDRKQAVCSNGHIWLRCFLTYQSCQSLIYRRCLLHDSIARHPAPEDPDWIKRLLQSPCPFCDSPVF
Specifications

Specifications

NP_036336
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DDRTARVLIGHISKKMNKQTFPEHCSLCKEILPFTDRKQAVCSNGHIWLRCFLTYQSCQSLIYRRCLLHDSIARHPAPEDPDWIKRLLQSPCPFCDSPVF
RUO
GTF3C4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9329
GTF3C4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ21002/KAT12/MGC138450/TFIII90/TFIIIC90/TFIIICdelta/TFiiiC2-90
GTF3C4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers