missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRK4 Partial ORF (NP_892027, 36 a.a. - 145 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002868-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188854

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating its deactivation. This gene has been linked to both genetic and acquired hypertension. [provided by RefSeq]

Sequence: LPPVSQCSELRHSIEKDYSSLCDKQPIGRRLFRQFCDTKPTLKRHIEFLDAVAEYEVADDEDRSDCGLSILDRFFNDKLAAPLPEIPPDVVTECRLGLKEENPSKKAFEE
Specifications

Specifications

NP_892027
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LPPVSQCSELRHSIEKDYSSLCDKQPIGRRLFRQFCDTKPTLKRHIEFLDAVAEYEVADDEDRSDCGLSILDRFFNDKLAAPLPEIPPDVVTECRLGLKEENPSKKAFEE
RUO
GRK4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2868
GRK4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GPRK2L/GPRK4/GRK4a/IT11
GRK4
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers