Learn More
Abnova™ Human GRIK3 Partial ORF (NP_000822, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002899-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics. [provided by RefSeq]
Sequence: NLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGMSpecifications
NP_000822 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGM | |
RUO | |
GRIK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2899 | |
GRIK3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EAA5/GLR7/GLUR7/GluR7a | |
GRIK3 | |
Recombinant | |
wheat germ expression system |