missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GRB10 Full-length ORF (NP_001001550.1, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002887-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188427

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene belongs to a small family of adapter proteins that are known to interact with a number of receptor tyrosine kinases and signaling molecules. This gene encodes a growth factor receptor-binding protein that interacts with insulin receptors and insulin-like growth-factor receptors. Overexpression of some isoforms of the encoded protein inhibits tyrosine kinase activity and results in growth suppression. This gene is imprinted in a highly isoform- and tissue-specific manner. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MNASLESLYSACSMQSDTVPLLQNGQHARSQPRASGPPRSIQPQVSPRQRVQRSQPVHILAVRRLQEEDQQFRTSSLPAIPNPFPELCGPGSPPVLTPGSLPPSQAAAKQDVKVFSEDGTSKVVEILADMTARDLCQLLVYKSHCVDDNSWTLVEHHPHLGLERCLEDHELVVQVESTMASESKFLFRKNYAKYEFFKNPMNFFPEQMVTWCQQSNGSQTQLLQNFLNSSSCPEIQGFLHVKELGKKSWKKLYVCLRRSGLYCSTKGTSKEPRHLQLLADLEDSNIFSLIAGRKQYNAPTDHGLCIKPNKVRNETKELRLLCAEDEQTRTCWMTAFRLLKYGMLLYQNYRIPQQRKALLSPFSTPVRSVSENSLVAMDFSGQTGRVIENPAEAQSAALEEGHAWRKRSTRMNILGSQSPLHPSTLSTVIHRTQHWFHGRISREESHRIIKQQGLVDGLFLLRDSQSNPKAFVLTLCHHQKIKNFQILPCEDDGQTFFSLDDGNTKFSDLIQLVDFYQLNKGVLPCKLKHHCIRVAL
Specifications

Specifications

NP_001001550.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GRB-IR/Grb-10/IRBP/KIAA0207/MEG1/RSS
GRB10
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2887
GRB10 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNASLESLYSACSMQSDTVPLLQNGQHARSQPRASGPPRSIQPQVSPRQRVQRSQPVHILAVRRLQEEDQQFRTSSLPAIPNPFPELCGPGSPPVLTPGSLPPSQAAAKQDVKVFSEDGTSKVVEILADMTARDLCQLLVYKSHCVDDNSWTLVEHHPHLGLERCLEDHELVVQVESTMASESKFLFRKNYAKYEFFKNPMNFFPEQMVTWCQQSNGSQTQLLQNFLNSSSCPEIQGFLHVKELGKKSWKKLYVCLRRSGLYCSTKGTSKEPRHLQLLADLEDSNIFSLIAGRKQYNAPTDHGLCIKPNKVRNETKELRLLCAEDEQTRTCWMTAFRLLKYGMLLYQNYRIPQQRKALLSPFSTPVRSVSENSLVAMDFSGQTGRVIENPAEAQSAALEEGHAWRKRSTRMNILGSQSPLHPSTLSTVIHRTQHWFHGRISREESHRIIKQQGLVDGLFLLRDSQSNPKAFVLTLCHHQKIKNFQILPCEDDGQTFFSLDDGNTKFSDLIQLVDFYQLNKGVLPCKLKHHCIRVAL
RUO
GRB10
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers