Learn More
Abnova™ Human GPX5 Full-length ORF (NP_003987.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002880-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the glutathione peroxidase family. It is specifically expressed in the epididymis in the mammalian male reproductive tract, and is androgen-regulated. Unlike mRNAs for other characterized glutathione peroxidases, this mRNA does not contain a selenocysteine (UGA) codon. Thus, the encoded protein is selenium-independent, and has been proposed to play a role in protecting the membranes of spermatozoa from the damaging effects of lipid peroxidation and/or preventing premature acrosome reaction. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MTTQLRVVHLLPLLLACFVQTSPKQEKMKMDCHKDEKGTIYDYEAIALNKNEYVSFKQYVGKHILFVNVATYCGLTAQYPGMSVQGEDLYLVSSFLRKGMSpecifications
NP_003987.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GPX5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2880 | |
GPX5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTTQLRVVHLLPLLLACFVQTSPKQEKMKMDCHKDEKGTIYDYEAIALNKNEYVSFKQYVGKHILFVNVATYCGLTAQYPGMSVQGEDLYLVSSFLRKGM | |
RUO | |
GPX5 | |
Recombinant | |
wheat germ expression system |