Learn More
Abnova™ Human GPR158 Partial ORF (NP_065803.1, 23 a.a. - 131 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand:  Abnova™ H00057512-Q01.10ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16129146
  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
Description
Sequence: RDPQGRPDSPRERTPKGKPHAQQPGRASASDSSAPWSRSTDGTILAQKLAEEVPMDVASYLYTGDSHQLKRANCSGRYELAGLPGKWPALASAHPSLHRALDTLTHATN
Specifications
Specifications
NP_065803.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RDPQGRPDSPRERTPKGKPHAQQPGRASASDSSAPWSRSTDGTILAQKLAEEVPMDVASYLYTGDSHQLKRANCSGRYELAGLPGKWPALASAHPSLHRALDTLTHATN
RUO
GPR158
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57512
GPR158 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ37801/KIAA1136/RP11-59G22.1
GPR158
Recombinant
wheat germ expression system