Learn More
Abnova™ Human GPR132 Partial ORF (NP_037477.1, 281 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029933-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subfamily member of the G-protein couple receptor (GPCR) superfamily. The encoded protein is a high-affinity receptor for lysophosphatidylcholine (LPC), a major phospholipid component of oxidized low density lipoprotein. This protein may react to LPC levels at sites of inflammation to limit the expansion of tissue-infiltrating cells. A similar protein in mouse is involved in cell cycle progression. [provided by RefSeq]
Sequence: GLEERLYTASVVFLCLSTVNGVADPIIYVLATDHSRQEVSRIHKGWKEWSMKTDVTRLTHSRDTEELQSPVALADHYTFSRPVHPPGSPCPAKRLIEESCSpecifications
NP_037477.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GLEERLYTASVVFLCLSTVNGVADPIIYVLATDHSRQEVSRIHKGWKEWSMKTDVTRLTHSRDTEELQSPVALADHYTFSRPVHPPGSPCPAKRLIEESC | |
RUO | |
GPR132 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29933 | |
GPR132 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
G2A/MGC99642 | |
GPR132 | |
Recombinant | |
wheat germ expression system |