Learn More
Abnova™ Human GP1BA Full-length ORF (AAH27955.1, 1 a.a. - 626 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002811-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Glycoprotein Ib (GP Ib) is a platelet surface membrane glycoprotein composed of a heterodimer, an alpha chain and a beta chain, that are linked by disulfide bonds. The Gp Ib functions as a receptor for von Willebrand factor (VWF). The complete receptor complex includes noncovalent association of the alpha and beta subunits with platelet glycoprotein IX and platelet glycoprotein V. The binding of the GP Ib-IX-V complex to VWF facilitates initial platelet adhesion to vascular subendothelium after vascular injury, and also initiates signaling events within the platelet that lead to enhanced platelet activation, thrombosis, and hemostasis. This gene encodes the alpha subunit. Several polymorphisms and mutations have been described in this gene, some of which are the cause of Bernard-Soulier syndromes and platelet-type von Willebrand disease. [provided by RefSeq]
Sequence: MPLLLLLLLLPSPLHPHPICEVSKVASHLEVNCDKRNLTALPPDLPKDTTILHLSENLLYTFSLATLMPYTRLTQLNLDRCELTKLQVDGTLPVLGTLDLSHNQLQSLPLLGQTLPALTVLDVSFNRLTSLPLGALRGLGELQELYLKGNELKTLPPGLLTPTPKLEKLSLANNNLTELPAGLLNGLENLDTLLLQENSLYTIPKGFFGSHLLPFAFLHGNPWLCNCEILYFRRWLQDNAENVYVWKQGVDVKAMTSNVASVQCDNSDKFPVYKYPGKGCPTLGDEGDTDLYDYYPEEDTEGDKVRATRTVVKFPTKAHTTPWGLFYSWSTASLDSQMPSSLHPTQESTKEQTTFPPRWTPNFTLHMESITFSKTPKSTTEPTPSPTTSEPVPEPAPNMTTLEPTPSPTTPEPTSEPAPSPTTPEPTPIPTIATSPTILVSATSLITPKSTFLTTTKPVSLLESTKKTIPELDQPPKLRGVLQGHLESSRNDPFLHPDFCCLLPLGFYVLGLFWLLFASVVLILLLSWVGHVKPQALDSGQGAALTTATQTTHLELQRGRQVTVPRAWLLFLRGSLPTFRSSLFLWVRPNGRLGPLVAGRRPSALSQGRGQDLLSTVSIRYSGHSLSpecifications
AAH27955.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BSS/CD42B/CD42b-alpha/GP1B/MGC34595 | |
GP1BA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2811 | |
GP1BA (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPLLLLLLLLPSPLHPHPICEVSKVASHLEVNCDKRNLTALPPDLPKDTTILHLSENLLYTFSLATLMPYTRLTQLNLDRCELTKLQVDGTLPVLGTLDLSHNQLQSLPLLGQTLPALTVLDVSFNRLTSLPLGALRGLGELQELYLKGNELKTLPPGLLTPTPKLEKLSLANNNLTELPAGLLNGLENLDTLLLQENSLYTIPKGFFGSHLLPFAFLHGNPWLCNCEILYFRRWLQDNAENVYVWKQGVDVKAMTSNVASVQCDNSDKFPVYKYPGKGCPTLGDEGDTDLYDYYPEEDTEGDKVRATRTVVKFPTKAHTTPWGLFYSWSTASLDSQMPSSLHPTQESTKEQTTFPPRWTPNFTLHMESITFSKTPKSTTEPTPSPTTSEPVPEPAPNMTTLEPTPSPTTPEPTSEPAPSPTTPEPTPIPTIATSPTILVSATSLITPKSTFLTTTKPVSLLESTKKTIPELDQPPKLRGVLQGHLESSRNDPFLHPDFCCLLPLGFYVLGLFWLLFASVVLILLLSWVGHVKPQALDSGQGAALTTATQTTHLELQRGRQVTVPRAWLLFLRGSLPTFRSSLFLWVRPNGRLGPLVAGRRPSALSQGRGQDLLSTVSIRYSGHSL | |
RUO | |
GP1BA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |