missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GOLPH3L Full-length ORF (NP_060648.2, 1 a.a. - 285 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055204-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121273

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is localized at the Golgi stack and may have a regulatory role in Golgi trafficking. [provided by RefSeq]

Sequence: MTTLTHRARRTEISKNSEKKMESEEDSNWEKSPDNEDSGDSKDIRLTLMEEVLLLGLKDKEGYTSFWNDCISSGLRGGILIELAMRGRIYLEPPTMRKKRLLDRKVLLKSDSPTGDVLLDETLKHIKATEPTETVQTWIELLTGETWNPFKLQYQLRNVRERIAKNLVEKGILTTEKQNFLLFDMTTHPVTNTTEKQRLVKKLQDSVLERWVNDPQRMDKRTLALLVLAHSSDVLENVFSSLTDDKYDVAMNRAKDLVELDPEVEGTKPSATEMIWAVLAAFNKS
Specifications

Specifications

NP_060648.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10687/GPP34R
GOLPH3L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55204
GOLPH3L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTTLTHRARRTEISKNSEKKMESEEDSNWEKSPDNEDSGDSKDIRLTLMEEVLLLGLKDKEGYTSFWNDCISSGLRGGILIELAMRGRIYLEPPTMRKKRLLDRKVLLKSDSPTGDVLLDETLKHIKATEPTETVQTWIELLTGETWNPFKLQYQLRNVRERIAKNLVEKGILTTEKQNFLLFDMTTHPVTNTTEKQRLVKKLQDSVLERWVNDPQRMDKRTLALLVLAHSSDVLENVFSSLTDDKYDVAMNRAKDLVELDPEVEGTKPSATEMIWAVLAAFNKS
RUO
GOLPH3L
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers