Learn More
Invitrogen™ Human GM130 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP296
Additional Details : Weight : 0.99000kg
Description
GM130/GOLGA2 Synthetic Peptide, PEP-296, from Invitrogen™.
GM130 is a putative cis-Golgi matrix protein that belongs to the golgin family of autoantigens possessing extensive coiled-coil domains. This protein interacts with GRASP65 and acts as a receptor for the vesicle docking protein p115 during vesicle fusion. This peptide corresponds to amino acids G(368) QVMESVRQLQMERDK (383) of human GM130 and can be used as a blocking peptide with polyclonal antibody PA1-077.Specifications
2801 | |
purified | |
Human GM130 | |
-20°C | |
Lyophilized | |
Blocking Assay, Control | |
GOLGA2 | |
No preservative | |
50 μg |
Human | |
Q08379 | |
1 mg/mL | |
Synthetic Peptide | |
water with no preservative | |
Golgin subfamily A member 2, 130 kDa cis-Golgi matrix protein, GM130, GM130 autoantigen, Golgin-95, GOLGA2 | |
Yes | |
Peptide | |
RUO - research use only |