Learn More
Abnova™ Human GLI1 Partial ORF (AAH13000.1, 401 a.a. - 500 a.a.) Recombinant Protein with GST-pstS1 tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002735-Q02.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GPLPRAPSISTVEPKREREGGPIREESRLTVPEGAMKPQPSPGAQSSCSSDHSPAGSAANTDSGVEMTGNAGGSTEDLSSLDEGPCIAGTGLSTLRRLENSpecifications
AAH13000.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.05kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
GPLPRAPSISTVEPKREREGGPIREESRLTVPEGAMKPQPSPGAQSSCSSDHSPAGSAANTDSGVEMTGNAGGSTEDLSSLDEGPCIAGTGLSTLRRLEN | |
RUO | |
GLI1 | |
Wheat Germ (in vitro) | |
GST-pstS1 | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2735 | |
GLI1 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLI | |
GLI1 | |
Recombinant | |
wheat germ expression system |