missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human GFR alpha-1 (aa 293-339) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Glial cell line-derived neurotrophic factor (GDNF) is a potent survival factor for central and peripheral neurons and is essential for the development of kidneys and the enteric nerves system. Physiological responses to GDNF require the presence of a novel glycosylphosphadidylinositol linked protein GDNFRalpha, which is a cell surface receptor for GDNF. The cDNAs encoding GDNFRalpha from human, rat, chicken and mouse have been cloned recently. GDNFRalpha was also termed Ret ligand 1 (RETL1) or TGF-beta-related neurotrophic factor receptor 1 (TrnR1) and nominated as GFR-a-1 recently. GFR-a-1 binds GDNF specifically and mediates activation of the Ret protein tyrosine kinase (PTK). Thus, GDNF, GFR-a and the Ret PTK form a complex to transduce GDNF signal and to mediate GDNF function.
Specifications
Specifications
| Accession Number | P56159 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 2674 |
| Name | Human GFR alpha-1 (aa 293-339) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AU042498; DKFZp313E1012; DKFZp686J0156; FLJ10561; FLJ31546; GDNF family receptor alpha 1; GDNF family receptor alpha-1; GDNF receptor alpha; GDNF receptor alpha-1; GDNFR; GDNFRA; GDNFR-alpha; GDNFR-alpha-1; gfr alpha 1; GFR alpha-1; GFRA1; GFR-alpha-1; glial cell line derived neurotrophic factor family receptor alpha 1; Glial cell line-derived neurotrophic factor receptor alpha; GPI-linked anchor protein; MGC23045; PI-linked cell-surface accessory protein; RET ligand 1; RET1L; RETL1; TGF-beta-related neurotrophic factor receptor 1; TRNR1 |
| Common Name | GFR alpha-1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction