Learn More
Abnova™ Human GEMIN4 Partial ORF (NP_056536, 959 a.a. - 1057 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00050628-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies (gems). The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]
Sequence: AAGPWVQGPEQDLTQEALFVYTQVFCHALHIMAMLHPEVCEPLYVLALETLTCYETLSKTNPSVSSLLQRAHEQRFLKSIAEGIGPEERRQTLLQKMSSSpecifications
NP_056536 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AAGPWVQGPEQDLTQEALFVYTQVFCHALHIMAMLHPEVCEPLYVLALETLTCYETLSKTNPSVSSLLQRAHEQRFLKSIAEGIGPEERRQTLLQKMSS | |
RUO | |
GEMIN4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
50628 | |
GEMIN4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434B131/DKFZp434D174/HC56/HCAP1/HHRF-1/p97 | |
GEMIN4 | |
Recombinant | |
wheat germ expression system |