missing translation for 'onlineSavingsMsg'
Learn More

Human GDNF Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-5380099
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
Product Code. Quantity unitSize
18732607 25 μg 25µg
18713889 100 μg 100µg
18704047 500 μg 500µg
3 options
This item is not returnable. View return policy

Product Code. 18732607

Brand: R&D Systems MAB212SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

GDNF Monoclonal specifically detects GDNF in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDNF
Applications Western Blot, ELISA
Classification Monoclonal
Clone 27106
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P39905
Gene Alias Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Gene Symbols GDNF
Host Species Mouse
Immunogen E. coli-derived recombinant human GDNF Arg109-Ile211 Accession # P39905
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2668
Test Specificity Detects human GDNF in ELISAs and Western blots. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) BDNF, rhCNTF, rhNeurturin, rh beta-NGF, rhNT-3, or rhNT-4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.