Learn More
Abnova™ Human GCDH Full-length ORF (NP_000150.1, 1 a.a. - 438 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002639-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the acyl-CoA dehydrogenase family. It catalyzes the oxidative decarboxylation of glutaryl-CoA to crotonyl-CoA and CO(2) in the degradative pathway of L-lysine, L-hydroxylysine, and L-tryptophan metabolism. It uses electron transfer flavoprotein as its electron acceptor. The enzyme exists in the mitochondrial matrix as a homotetramer of 45-kD subunits. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASKSpecifications
NP_000150.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACAD5/GCD | |
GCDH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2639 | |
GCDH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASK | |
RUO | |
GCDH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |