missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GAS2 Partial ORF (NP_005247, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002620-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135331

  • 6245.00 NOK / 25µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

The protein encoded by this gene is a caspase-3 substrate that plays a role in regulating microfilament and cell shape changes during apoptosis. It can also modulate cell susceptibility to p53-dependent apoptosis by inhibiting calpain activity. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MCTALSPKVRSGPGLSDMHQYSQWLASRHEANLLPMKEDLALWLTNLLGKEITAETFMEKLDNGALLCQLAETMQEKFKESMDANKPTKNLPLKKIPCKTSAPSGSFFAR
Specifications

Specifications

NP_005247
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MCTALSPKVRSGPGLSDMHQYSQWLASRHEANLLPMKEDLALWLTNLLGKEITAETFMEKLDNGALLCQLAETMQEKFKESMDANKPTKNLPLKKIPCKTSAPSGSFFAR
RUO
GAS2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2620
GAS2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC32610
GAS2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers