Learn More
Abnova™ Human GALNT3 Partial ORF (NP_004473.1, 43 a.a. - 138 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002591-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein encoded by this gene is highly homologous to other family members, however the enzymes have different substrate specificities. [provided by RefSeq]
Sequence: VQYSKEESRMERNMKNKNKMLDLMLEAVNNIKDAMPKMQIGAPVRQNIDAGERPCLQGYYTAAELKPVLDRPPQDSNAPGASGKAFKTTNLSVEEQSpecifications
NP_004473.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.3kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VQYSKEESRMERNMKNKNKMLDLMLEAVNNIKDAMPKMQIGAPVRQNIDAGERPCLQGYYTAAELKPVLDRPPQDSNAPGASGKAFKTTNLSVEEQ | |
RUO | |
GALNT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2591 | |
GALNT3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686C10199/GalNAc-T3/HFTC/HHS/MGC61909 | |
GALNT3 | |
Recombinant | |
wheat germ expression system |