Learn More
Abnova™ Human Gage12B Full-Length Orf (Adr82812.1, 1 A.A.-117 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
Brand: Abnova™ H00729428-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQCQDPAAAQEGEDEGASAGQGPKPEAHSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEGEKQSQC
Specifications
ADR82812.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
GAGE12B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQCQDPAAAQEGEDEGASAGQGPKPEAHSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEGEKQSQC | |
RUO | |
GAGE12B | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
729428 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC163246 | |
GAGE12B | |
Liquid |
For Research Use Only