Learn More
Abnova™ Human GABRE Partial ORF (AAH59376.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002564-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the ligand-gated ionic channel (TC 1.A.9) family. It encodes the gamma-aminobutyric acid (GABA) A receptor which is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes an epsilon subunit. It is mapped to chromosome Xq28 in a cluster comprised of genes encoding alpha 3, beta 4 and theta subunits of the same receptor. Alternatively spliced transcript variants have been identified, but only one is thought to encode a protein. [provided by RefSeq]
Sequence: MLSKVLPVFLGILLILQSRVEGPQTESKNEASSRDVVYGPQPQPLENQLLSEETKSTETETGSRVGKLPEASRILNTILSNYDHKLRPGIGEKPTVVTVESpecifications
AAH59376.1 | |
Liquid | |
2564 | |
GABRE (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GABRE | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MLSKVLPVFLGILLILQSRVEGPQTESKNEASSRDVVYGPQPQPLENQLLSEETKSTETETGSRVGKLPEASRILNTILSNYDHKLRPGIGEKPTVVTVE | |
RUO | |
GABRE | |
Yes | |
wheat germ expression system |