missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FSCB Full-length ORF (NP_115511.2, 1 a.a. - 825 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084075-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125613

  • 5540.00 NOK / 25µg
Estimated Shipment: 16-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MVGKSQQTDVIEKKKHMAIPKSSSPKATHRIGNTSGSKGSYSAKAYESIRVSSELQQTWTKRKHGQEMTSKSLQTDTIVEEKKEVKLVEETVVPEEKSADVREAAIELPESVQDVEIPPNIPSVQLKMDRSQQTSRTGYWTMMNIPPVEKVDKEQQTYFSESEIVVISRPDSSSTKSKEDALKHKSSGKIFASEQPEFQPATNSNEEIGQKNISRTSFTQETKKGPPVLLEDELREEVTVPVVQEGSAVKKVASAEIEPPSTEKFPAKIQPPLVEEATAKAEPRPAEETHVQVQPSTEETPDAEAATAVAENSVKVQPPPAEEAPLVEFPAEIQPPSAEESPSVELLAEILPPSAEESLSEEPPAEILPPPAEKSPSVEPLGEIRSPSAQKAPIEVQPLPAEGALEEASAKVEPPTVEETLADVQPLLPEEAPREEARELQLSTAMETPAEEAPTEFQSPLPKETTAEEASAEIQLLAATEPPADETPAEARSPLSEETSAEEAHAEVQSPLAEETTAEEASAEIQLLAAIEAPADETPAEAQSPLSEETSAEEAPAEVQSPSAKGVSIEEAPLELQPPSGEETTAEEASAAIQLLAATEASAEEAPAEVQPPPAEEAPAEVQPPPAEEAPAEVQPPPAEETPAEVQPPPAEEAPAEVQPPPAEEAPAEVQPPPAEEAPAEVQSLPAEETPIEETLAAVHSPPADDVPAEEASVDKHSPPADLLLTEEFPIGEASAEVSPPPSEQTPEDEALVENVSTEFQSPQVAGIPAVKLGSVVLEGEAKFEEVSKINSVLKDLSNTNDGQAPTLEIESVFHIELKQRPPEL
Specifications

Specifications

NP_115511.2
Liquid
84075
FSCB (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVGKSQQTDVIEKKKHMAIPKSSSPKATHRIGNTSGSKGSYSAKAYESIRVSSELQQTWTKRKHGQEMTSKSLQTDTIVEEKKEVKLVEETVVPEEKSADVREAAIELPESVQDVEIPPNIPSVQLKMDRSQQTSRTGYWTMMNIPPVEKVDKEQQTYFSESEIVVISRPDSSSTKSKEDALKHKSSGKIFASEQPEFQPATNSNEEIGQKNISRTSFTQETKKGPPVLLEDELREEVTVPVVQEGSAVKKVASAEIEPPSTEKFPAKIQPPLVEEATAKAEPRPAEETHVQVQPSTEETPDAEAATAVAENSVKVQPPPAEEAPLVEFPAEIQPPSAEESPSVELLAEILPPSAEESLSEEPPAEILPPPAEKSPSVEPLGEIRSPSAQKAPIEVQPLPAEGALEEASAKVEPPTVEETLADVQPLLPEEAPREEARELQLSTAMETPAEEAPTEFQSPLPKETTAEEASAEIQLLAATEPPADETPAEARSPLSEETSAEEAHAEVQSPLAEETTAEEASAEIQLLAAIEAPADETPAEAQSPLSEETSAEEAPAEVQSPSAKGVSIEEAPLELQPPSGEETTAEEASAAIQLLAATEASAEEAPAEVQPPPAEEAPAEVQPPPAEEAPAEVQPPPAEETPAEVQPPPAEEAPAEVQPPPAEEAPAEVQPPPAEEAPAEVQSLPAEETPIEETLAAVHSPPADDVPAEEASVDKHSPPADLLLTEEFPIGEASAEVSPPPSEQTPEDEALVENVSTEFQSPQVAGIPAVKLGSVVLEGEAKFEEVSKINSVLKDLSNTNDGQAPTLEIESVFHIELKQRPPEL
RUO
FSCB
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
114.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C14orf155/DKFZp434F1017/DKFZp686A1639/DKFZp686J0539
FSCB
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers