missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FOXM1 Full-length ORF (ADZ15712.1, 1 a.a. - 748 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002305-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MKTSPRRPLILKRRRLPLPVQNAPSETSEEEPKRSPAQQESNQAEASKEVAESNSCKFPAGIKIINHPTMPNTQVVAIPNNANIHSIITALTAKGKESGSSGPNKFILISCGGAPTQPPGLRPQTQTSYDAKRTEVTLETLGPKPAARDVNLPRPPGALCEQKRETCADGEAAGCTINNSLSNIQWLRKMSSDGLGSRSIKQEMEEKENCHLEQRQVKVEEPSRPSASWQNSVSERPPYSYMAMIQFAINSTERKRMTLKDIYTWIEDHFPYFKHIAKPGWKNSIRHNLSLHDMFVRETSANGKVSFWTIHPSANRYLTLDQVFKQQKRPNPELRRNMTIKTELPLGARRKMKPLLPRVSSYLVPIQFPVNQSLVLQPSVKVPLPLAASLMSSELARHSKRVRIAPKVLLAEEGIAPLSSAGPGKEEKLLFGEGFSPLLPVQTIKEEEIQPGEEMPHLARPIKVESPPLEEWPSPAPSFKEESSHSWEDSSQSPTPRPKKSYSGLRSPTRCVSEMLVIQHRERRERSRSRRKQHLLPPCVDEPELLFSEGPSTSRWAAELPFPADSSDPASQLSYSQEVGGPFKTPIKETLPISSTPSKSVLPRTPESWRLTPPAKVGGLDFSPVQTPQGASDPLPDPLGLMDLSTTPLQSAPPLESPQRLLSSEPLDLISVPFGNSSPSDIDVPKPGSPEPQVSGLAANRSLTEGLVLDTMNDSLSKILLDISFPGLDEDPLGPDNINWSQFIPELQSpecifications
ADZ15712.1 | |
Liquid | |
2305 | |
FOXM1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MKTSPRRPLILKRRRLPLPVQNAPSETSEEEPKRSPAQQESNQAEASKEVAESNSCKFPAGIKIINHPTMPNTQVVAIPNNANIHSIITALTAKGKESGSSGPNKFILISCGGAPTQPPGLRPQTQTSYDAKRTEVTLETLGPKPAARDVNLPRPPGALCEQKRETCADGEAAGCTINNSLSNIQWLRKMSSDGLGSRSIKQEMEEKENCHLEQRQVKVEEPSRPSASWQNSVSERPPYSYMAMIQFAINSTERKRMTLKDIYTWIEDHFPYFKHIAKPGWKNSIRHNLSLHDMFVRETSANGKVSFWTIHPSANRYLTLDQVFKQQKRPNPELRRNMTIKTELPLGARRKMKPLLPRVSSYLVPIQFPVNQSLVLQPSVKVPLPLAASLMSSELARHSKRVRIAPKVLLAEEGIAPLSSAGPGKEEKLLFGEGFSPLLPVQTIKEEEIQPGEEMPHLARPIKVESPPLEEWPSPAPSFKEESSHSWEDSSQSPTPRPKKSYSGLRSPTRCVSEMLVIQHRERRERSRSRRKQHLLPPCVDEPELLFSEGPSTSRWAAELPFPADSSDPASQLSYSQEVGGPFKTPIKETLPISSTPSKSVLPRTPESWRLTPPAKVGGLDFSPVQTPQGASDPLPDPLGLMDLSTTPLQSAPPLESPQRLLSSEPLDLISVPFGNSSPSDIDVPKPGSPEPQVSGLAANRSLTEGLVLDTMNDSLSKILLDISFPGLDEDPLGPDNINWSQFIPELQ | |
RUO | |
FOXM1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHL16/FOXM1B/HFH-11/HFH11/HNF-3/INS-1/MPHOSPH2/MPP-2/MPP2/PIG29/TGT3/TRIDENT | |
FOXM1 | |
Yes | |
wheat germ expression system |