missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FLJ36180 Partial ORF (NP_848651, 369 a.a. - 468 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00339976-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122817

  • 6245.00 NOK / 25µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: PKPPGDLFSLIGLKIGDDYSLWVSSPLKGQHVREPVCKVGVFLDYESGHIAFYNGTDESLIYSFPQASFQEALRPIFSPCLPNEGTNTDPLTICSLNSHV
Specifications

Specifications

NP_848651
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PKPPGDLFSLIGLKIGDDYSLWVSSPLKGQHVREPVCKVGVFLDYESGHIAFYNGTDESLIYSFPQASFQEALRPIFSPCLPNEGTNTDPLTICSLNSHV
RUO
TRIML1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
339976
FLJ36180 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ36180/MGC138638/MGC138639/RNF209
TRIML1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers