missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FLCN Partial ORF (NP_653207, 198 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00201163-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122407

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is located within the Smith-Magenis syndrome region on chromosome 17. Mutations in this gene are associated with Birt-Hogg-Dube syndrome, which is characterized by fibrofolliculomas, renal tumors, lung cysts, and pneumothorax. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: DELQGKALKVFEAEQFGCPQRAQRMNTAFTPFLHQRNGNAARSLTSLTSDDNLWACLHTSFAWLLKACGSRLTEKLLEGAPTEDTLVQMEKLAGEAGVL
Specifications

Specifications

NP_653207
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DELQGKALKVFEAEQFGCPQRAQRMNTAFTPFLHQRNGNAARSLTSLTSDDNLWACLHTSFAWLLKACGSRLTEKLLEGAPTEDTLVQMEKLAGEAGVL
RUO
FLCN
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
201163
FLCN (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BHD/DKFZp547A118/FLCL/FLJ45004/FLJ99377/MGC17998/MGC23445
FLCN
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers