Learn More
Abnova™ Human FGF5 Full-length ORF (NP_149134.1, 1 a.a. - 123 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002250-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was identified as an oncogene, which confers transforming potential when transfected into mammalian cells. Targeted disruption of the homolog of this gene in mouse resulted in the phenotype of abnormally long hair, which suggested a function as an inhibitor of hair elongation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MSLSFLLLLFFSHLILSAWAHGEKRLAPKGQPGPAATDRNPRGSSSRQSSSSAMSSSSASSSPAASLGSQGSGLEQSSFQWSPSGRRTGSLYCRVGIGFHLQIYPDGKVNGSHEANMLSQVHRSpecifications
NP_149134.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBGF-5/Smag-82 | |
FGF5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2250 | |
FGF5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLSFLLLLFFSHLILSAWAHGEKRLAPKGQPGPAATDRNPRGSSSRQSSSSAMSSSSASSSPAASLGSQGSGLEQSSFQWSPSGRRTGSLYCRVGIGFHLQIYPDGKVNGSHEANMLSQVHR | |
RUO | |
FGF5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |