missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®

Product Code. 17876418
Click to view available options
Quantity:
2 μg
250 μg
50 μg
500 μg
Unit Size:
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17876418

Brand: Gibco™ AF1001950UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21781
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 18.9 kDa
Name Human FGF-7 (KGF), Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 50 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Common Name KGF
Gene Symbol FGF7
Biological Activity Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT
Content And Storage -20°C
Activity Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.