missing translation for 'onlineSavingsMsg'
Learn More

Human FGF-11 Antibody, R&D Systems™

Product Code. 18152281
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18152281 500 μg 500µg
18792959 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18152281 Supplier R&D Systems Supplier No. MAB1634

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-11 Monoclonal specifically detects FGF-11 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-11
Applications Western Blot
Classification Monoclonal
Clone 224812
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q2YDX8
Gene Alias FGF11, FGF-11, FHF-3, FHF3Fibroblast growth factor homologous factor 3, fibroblast growth factor 11, FLJ16061, MGC102953, MGC45269
Gene Symbols FGF11
Host Species Mouse
Immunogen E. coli-derived recombinant human FGF-11 Met1-Pro225 (predicted) Accession # Q2YDX8
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2256
Test Specificity Detects human FGF-11 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with bovine FGF acidic, bovine FGF basic, recombinant human (rh) FGF acidic, rhFGF basic, rhFGF-3, -4, -5, -6, -7, -8b, -9, -10, -12, -13, -16, -17, -18, -19, -20, recombinant mouse FGF-8b, 8c, or -15 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.