Learn More
Abnova™ Human FETUB Full-length ORF (AAH74734.1, 1 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026998-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the fetuin family, part of the cystatin superfamily of cysteine protease inhibitors. Fetuins have been implicated in several diverse functions, including osteogenesis and bone resorption, regulation of the insulin and hepatocyte growth factor receptors, and response to systemic inflammation. This protein may be secreted by cells. [provided by RefSeq]
Sequence: MGLLLPLAPCILVLCCGAMSPPQLALNPSALLSRGCNDSDVLAVAGFALRDINKDRKDGYVLRLNRVNDAQEYRRGGLGSLFYLTLDVLETDCHVLRKKAWQDCGMRIFFESVYGQCKAIFYMNNPSRVLYLAAYNCTLRPVSKKKIYMTCPDCPSSIPTDSSNHQVLEAATESLAKYKNENTSKQYSLFKVTRASSQWVVGPSYFVEYLIKESPCTKSQASSCSLQSSDSVPVGLCKGSLTRTHWEKFVSVTCDFFESQAPATGSENSAVNQKPTNLPKVEESQQKNTPPTDSPSKAGPRGSVQYLPDLDDKNSQEKGPQEAFPVHLDLTTNPQGETLDISFLFLEPMEEKLVVLPFPKEKARTAECPGPAQNASPLVLPPSpecifications
AAH74734.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
16G2/Gugu/IRL685 | |
FETUB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26998 | |
FETUB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGLLLPLAPCILVLCCGAMSPPQLALNPSALLSRGCNDSDVLAVAGFALRDINKDRKDGYVLRLNRVNDAQEYRRGGLGSLFYLTLDVLETDCHVLRKKAWQDCGMRIFFESVYGQCKAIFYMNNPSRVLYLAAYNCTLRPVSKKKIYMTCPDCPSSIPTDSSNHQVLEAATESLAKYKNENTSKQYSLFKVTRASSQWVVGPSYFVEYLIKESPCTKSQASSCSLQSSDSVPVGLCKGSLTRTHWEKFVSVTCDFFESQAPATGSENSAVNQKPTNLPKVEESQQKNTPPTDSPSKAGPRGSVQYLPDLDDKNSQEKGPQEAFPVHLDLTTNPQGETLDISFLFLEPMEEKLVVLPFPKEKARTAECPGPAQNASPLVLPP | |
RUO | |
FETUB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |