missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FCRL2 Partial ORF (NP_110391, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079368-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139996

  • 4130.00 NOK / 10µg
Estimated Shipment: 29-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein has four extracellular C2-type immunoglobulin domains, a transmembrane domain and a cytoplasmic domain that contains one immunoreceptor-tyrosine activation motif and two immunoreceptor-tyrosine inhibitory motifs. This protein may be a prognostic marker for chronic lymphocytic leukemia. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: MLLWSLLVIFDAVTEQADSLTLVAPSSVFEGDSIVLKCQGEQNWKIQKMAYHKDNKELSVFKKFSDFLIQSAVLSDSGNYFCSTKGQLFLWDKTSNIVKIKVQELFQRPV
Specifications

Specifications

NP_110391
Liquid
79368
FCRL2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FCRH2/IFGP4/IRTA4/SPAP1/SPAP1A/SPAP1B/SPAP1C
FCRL2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLLWSLLVIFDAVTEQADSLTLVAPSSVFEGDSIVLKCQGEQNWKIQKMAYHKDNKELSVFKKFSDFLIQSAVLSDSGNYFCSTKGQLFLWDKTSNIVKIKVQELFQRPV
RUO
FCRL2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers