Learn More
Abnova™ Human FBLN1 Partial ORF (AAH22497, 151 a.a. - 250 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002192-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Fibulin 1 is a secreted glycoprotein that becomes incorporated into a fibrillar extracellular matrix. Calcium-binding is apparently required to mediate its binding to laminin and nidogen. It mediates platelet adhesion via binding fibrinogen. Four splice variants which differ in the 3' end have been identified. Each variant encodes a different isoform, but no functional distinctions have been identified among the four variants. [provided by RefSeq]
Sequence: GDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRDTGDEVVCSCFVGYQLLSDGVSCEDVNECITGSHSCRLGESCINTVGSFRCQRDSSCGTSpecifications
AAH22497 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GDLDVGGLQETDKIIEVEEEQEDPYLNDRCRGGGPCKQQCRDTGDEVVCSCFVGYQLLSDGVSCEDVNECITGSHSCRLGESCINTVGSFRCQRDSSCGT | |
RUO | |
FBLN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2192 | |
FBLN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBLN | |
FBLN1 | |
Recombinant | |
wheat germ expression system |