Learn More
Abnova™ Human FARP1 Full-length ORF (NP_001001715.1, 1 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010160-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene was originally isolated through subtractive hybridization due to its increased expression in differentiated chondrocytes versus dedifferentiated chondrocytes. The resulting protein contains a predicted ezrin-like domain, a Dbl homology domain, and a pleckstrin homology domain. It is believed to be a member of the band 4.1 superfamily whose members link the cytoskeleton to the cell membrane. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: MGEIEQRPTPGSRLGAPENSGISTLERGQKPPPTPSGKLVSIKIQMLDDTQEAFEVPMVSSSSFLKAIGSSWTGWVLRCSMKPKHHSHLIEKFGEDRILTHLTGSISYTNWAGSRSLAVTVTEELLNLFSpecifications
NP_001001715.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDEP/MGC87400/PLEKHC2 | |
FARP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10160 | |
FARP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGEIEQRPTPGSRLGAPENSGISTLERGQKPPPTPSGKLVSIKIQMLDDTQEAFEVPMVSSSSFLKAIGSSWTGWVLRCSMKPKHHSHLIEKFGEDRILTHLTGSISYTNWAGSRSLAVTVTEELLNLF | |
RUO | |
FARP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |