missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FAM3B Full-length ORF (AAH57829.1, 1 a.a. - 235 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054097-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120463

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MRPLAGGLLKVVFVVFASLCAWYSGYLLAELIPDAPLSSAAYSIRSIGERPVLKAPVPKRQKCDHWTPCPSDTYAYRLLSGGGRSKYAKICFEDNLLMGEQLGNVARGINIAIVNYVTGNVTATRCFDMYEGDNSGPMTKFIQSAAPKSLLFMVTYDDGSTRLNNDAKNAIEALGSKEIRNMKFRSSWVFIAAKGLELPSEIQREKINHSDAKNNRYSGWPAEIQIEGCIPKERS
Specifications

Specifications

AAH57829.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.59kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2-21/C21orf11/C21orf76/ORF9/PANDER/PRED44
FAM3B
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54097
FAM3B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRPLAGGLLKVVFVVFASLCAWYSGYLLAELIPDAPLSSAAYSIRSIGERPVLKAPVPKRQKCDHWTPCPSDTYAYRLLSGGGRSKYAKICFEDNLLMGEQLGNVARGINIAIVNYVTGNVTATRCFDMYEGDNSGPMTKFIQSAAPKSLLFMVTYDDGSTRLNNDAKNAIEALGSKEIRNMKFRSSWVFIAAKGLELPSEIQREKINHSDAKNNRYSGWPAEIQIEGCIPKERS
RUO
FAM3B
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers