missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FAM29A Full-length ORF (NP_060115.3, 1 a.a. - 955 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054801-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140763

  • 5420.23 NOK / 25µg

Estimated Shipment: 10-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MSSASVTAFEKEHLWMYLQALGFEPGPATIACGKIVSHTHLGVNMFDKLNRDAFHIISYFLFQVLDQSLTKEVFKFCWPPFDQKSDTEFRKHCCEWIKRISGECGSSFPQVVGSLFLSPGGPKFIHLMYHFARFVAMKYIKSNSKNSSHHFVETFNIKPQDLHKCIARCHFARSRFLQILQRQDCVTQKYQENAQLSVKQVRNLRSECIGLENQIKKMEPYDDHSNMEEKIQKVRSLWASVNETLMFLEKEREVVSSVLSLVNQYALDGTNVAINIPRLLLDKIEKQMFQLHIGNVYEAGKLNLLTVIQLLNEVLKVMKYERCQADQARLTVDLHYLEKETKFQKERLSDLKHMRYRIKDDLTTIRHSVVEKQGEWHKKWKEFLGLSPFSLIKGWTPSVDLLPPMSPLSFDPASEEVYAKSILCQYPASLPDAHKQHNQENGCRGDSDTLGALHDLANSPASFLSQSVSSSDRNSVTVLEKDTKMGTPKEKNEAISKKIPEFEVENSPLSDVAKNTESSAFGGSLPAKKSDPFQKEQDHLVEEVARAVLSDSPQLSEGKEIKLEELIDSLGSNPFLTRNQIPRTPENLITEIRSSWRKAIEMEENRTKEPIQMDAEHREVLPESLPVLHNQREFSMADFLLETTVSDFGQSHLTEEKVISDCECVPQKHVLTSHIDEPPTQNQSDLLNKKVICKQDLECLAFTKLSETSRMETFSPAVGNRIDVMGGSEEEFMKILDHLEVSCNKPSTNKTMLWNSFQISSGISSKSFKDNDFGILHETLPEEVGHLSFNSSSSSEANFKLEPNSPMHGGTLLEDVVGGRQTTPESDFNLQALRSRYEALKKSLSKKREESYLSNSQTPERHKPELSPTPQNVQTDDTLNFLDTCDLHTEHIKPSLRTSIGERKRSLSPLIKFSPVEQRLRTTIACSLGELPNLKEEDILNKSLDAKEPPSDLTR
Specifications

Specifications

NP_060115.3
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
135kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Dgt6/MGC102696/MGC138798/MGC138799/RP11-296P7.3
FAM29A
Yes
wheat germ expression system
Liquid
54801
FAM29A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSASVTAFEKEHLWMYLQALGFEPGPATIACGKIVSHTHLGVNMFDKLNRDAFHIISYFLFQVLDQSLTKEVFKFCWPPFDQKSDTEFRKHCCEWIKRISGECGSSFPQVVGSLFLSPGGPKFIHLMYHFARFVAMKYIKSNSKNSSHHFVETFNIKPQDLHKCIARCHFARSRFLQILQRQDCVTQKYQENAQLSVKQVRNLRSECIGLENQIKKMEPYDDHSNMEEKIQKVRSLWASVNETLMFLEKEREVVSSVLSLVNQYALDGTNVAINIPRLLLDKIEKQMFQLHIGNVYEAGKLNLLTVIQLLNEVLKVMKYERCQADQARLTVDLHYLEKETKFQKERLSDLKHMRYRIKDDLTTIRHSVVEKQGEWHKKWKEFLGLSPFSLIKGWTPSVDLLPPMSPLSFDPASEEVYAKSILCQYPASLPDAHKQHNQENGCRGDSDTLGALHDLANSPASFLSQSVSSSDRNSVTVLEKDTKMGTPKEKNEAISKKIPEFEVENSPLSDVAKNTESSAFGGSLPAKKSDPFQKEQDHLVEEVARAVLSDSPQLSEGKEIKLEELIDSLGSNPFLTRNQIPRTPENLITEIRSSWRKAIEMEENRTKEPIQMDAEHREVLPESLPVLHNQREFSMADFLLETTVSDFGQSHLTEEKVISDCECVPQKHVLTSHIDEPPTQNQSDLLNKKVICKQDLECLAFTKLSETSRMETFSPAVGNRIDVMGGSEEEFMKILDHLEVSCNKPSTNKTMLWNSFQISSGISSKSFKDNDFGILHETLPEEVGHLSFNSSSSSEANFKLEPNSPMHGGTLLEDVVGGRQTTPESDFNLQALRSRYEALKKSLSKKREESYLSNSQTPERHKPELSPTPQNVQTDDTLNFLDTCDLHTEHIKPSLRTSIGERKRSLSPLIKFSPVEQRLRTTIACSLGELPNLKEEDILNKSLDAKEPPSDLTR
RUO
FAM29A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates