missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FAM120B Full-length ORF (NP_115824.1, 1 a.a. - 910 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084498-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MGVRGLQGFVGSTCPHICTVVNFKELAEHHRSKYPGCTPTIVVDAMCCLRYWYTPESWICGGQWREYFSALRDFVKTFTAAGIKLIFFFDGMVEQDKRDEWVKRRLKNNREISRIFHYIKSHKEQPGRNMFFIPSGLAVFTRFALKTLGQETLCSLQEADYEVASYGLQHNCLGILGEDTDYLIYDTCPYFSISELCLESLDTVMLCREKLCESLGLCVADLPLLACLLGNDIIPEGMFESFRYKCLSSYTSVKENFDKKGNIILAVSDHISKVLYLYQGEKKLEEILPLGPNKALFYKGMASYLLPGQKSPWFFQKPKGVITLDKQVISTSSDAESREEVPMCSDAESRQEVPMCTGPESRREVPVYTDSEPRQEVPMCSDPEPRQEVPTCTGPESRREVPMCSDPEPRQEVPMCTGPEARQEVPMYTDSEPRQEVPMYTDSEPRQEVPMYTGSEPRQEVPMYTGPESRQEVPMYTGPESRQEVLIRTDPESRQEIMCTGHESKQEVPICTDPISKQEDSMCTHAEINQKLPVATDFEFKLEALMCTNPEIKQEDPTNVGPEVKQQVTMVSDTEILKVARTHHVQAESYLVYNIMSSGEIECSNTLEDELDQALPSQAFIYRPIRQRVYSLLLEDCQDVTSTCLAVKEWFVYPGNPLRHPDLVRPLQMTIPGGTPSLKILWLNQEPEIQVRRLDTLLACFNLSSSREELQAVESPFQALCCLLIYLFVQVDTLCLEDLHAFIAQALCLQGKSTSQLVNLQPDYINPRAVQLGSLLVRGLTTLVLVNSACGFPWKTSDFMPWNVFDGKLFHQKYLQSEKGYAVEVLLEQNRSRLTKFHNLKAVVCKACMKENRRITGRAHWGSHHAGRWGRQGSSYHRTGSGYSRSSQGQPWRDQGPGSRQYEHDQWRRYSpecifications
NP_115824.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
130.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCPG/FLJ55614/KIAA0183/KIAA1838/PGCC1/dJ894D12.1 | |
FAM120B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84498 | |
FAM120B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGVRGLQGFVGSTCPHICTVVNFKELAEHHRSKYPGCTPTIVVDAMCCLRYWYTPESWICGGQWREYFSALRDFVKTFTAAGIKLIFFFDGMVEQDKRDEWVKRRLKNNREISRIFHYIKSHKEQPGRNMFFIPSGLAVFTRFALKTLGQETLCSLQEADYEVASYGLQHNCLGILGEDTDYLIYDTCPYFSISELCLESLDTVMLCREKLCESLGLCVADLPLLACLLGNDIIPEGMFESFRYKCLSSYTSVKENFDKKGNIILAVSDHISKVLYLYQGEKKLEEILPLGPNKALFYKGMASYLLPGQKSPWFFQKPKGVITLDKQVISTSSDAESREEVPMCSDAESRQEVPMCTGPESRREVPVYTDSEPRQEVPMCSDPEPRQEVPTCTGPESRREVPMCSDPEPRQEVPMCTGPEARQEVPMYTDSEPRQEVPMYTDSEPRQEVPMYTGSEPRQEVPMYTGPESRQEVPMYTGPESRQEVLIRTDPESRQEIMCTGHESKQEVPICTDPISKQEDSMCTHAEINQKLPVATDFEFKLEALMCTNPEIKQEDPTNVGPEVKQQVTMVSDTEILKVARTHHVQAESYLVYNIMSSGEIECSNTLEDELDQALPSQAFIYRPIRQRVYSLLLEDCQDVTSTCLAVKEWFVYPGNPLRHPDLVRPLQMTIPGGTPSLKILWLNQEPEIQVRRLDTLLACFNLSSSREELQAVESPFQALCCLLIYLFVQVDTLCLEDLHAFIAQALCLQGKSTSQLVNLQPDYINPRAVQLGSLLVRGLTTLVLVNSACGFPWKTSDFMPWNVFDGKLFHQKYLQSEKGYAVEVLLEQNRSRLTKFHNLKAVVCKACMKENRRITGRAHWGSHHAGRWGRQGSSYHRTGSGYSRSSQGQPWRDQGPGSRQYEHDQWRRY | |
RUO | |
FAM120B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |