Learn More
Abnova™ Human EXTL1 Partial ORF (NP_004446, 141 a.a. - 241 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002134-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the multiple exostoses (EXT) family of glycosyltransferases, which function in the chain polymerization of heparan sulfate and heparin. The encoded protein harbors alpha 1,4- N-acetylglucosaminyltransferase activity, and is involved in chain elongation of heparan sulfate and possibly heparin. [provided by RefSeq]
Sequence: AQTGECSSMPLQWNRGRNHLVLRLHPAPCPRTFQLGQAMVAEASPTVDSFRPGFDVALPFLPEAHPLRGGAPGQLRQHSPQPGVALLALEEERGGWRTADTSpecifications
NP_004446 | |
Liquid | |
2134 | |
EXTL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EXTL/MGC70794 | |
EXTL1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AQTGECSSMPLQWNRGRNHLVLRLHPAPCPRTFQLGQAMVAEASPTVDSFRPGFDVALPFLPEAHPLRGGAPGQLRQHSPQPGVALLALEEERGGWRTADT | |
RUO | |
EXTL1 | |
Wheat Germ (in vitro) | |
GST |