missing translation for 'onlineSavingsMsg'
Learn More

Human ErbB3/Her3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. p-5317134
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18736458 25 μg 25µg
18721728 100 μg 100µg
18761289 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18736458 Supplier R&D Systems Supplier No. MAB348SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ErbB3/Her3 Monoclonal specifically detects ErbB3/Her3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ErbB3/Her3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 66219
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P21860
Gene Alias c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
Gene Symbols ERBB3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ErbB3 Ser20-Thr643 Accession # P21860
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2065
Test Specificity Detects human ErbB3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows no cross-reactivity with recombinant human (rh) EGF R, rhErbB4, or rhErbB2.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.