Learn More
Abnova™ Human EPYC Full-length ORF (AAH30958, 20 a.a. - 322 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001833-P02.25ug
Additional Details : Weight : 0.00010kg
Description
Dermatan sulfate proteoglycan 3 is a member of the small leucine-rich repeat proteoglycan family. This gene is composed of seven exons. It regulates fibrillogenesis by interacting with collagen fibrils and other extracellular matrix proteins. [provided by RefSeq]
Sequence: APTLESINYDSETYDATLEDLDNLYNYENIPVGKVEIEIATVMPSGNRELLTPPPQPEKAQEEEEEEESTPRLIDGSSPQEPEFTGVLGPHTNEDFPTCLLCTCISTTVYCDDHELDAIPPLPKNTAYFYSRFNRIKKINKNDFASLSDLKRIDLTSNLISEIDEDAFRKLPQLRELVLRDNKIRQLPELPTTLTFIDISNNRLGRKGIKQEAFKDMYDLHHLYLTDNNLDHIPLPLPENLRALHLQNNNILEMHEDTFCNVKNLTYIRKALEDIRLDGNPINLSKTPQAYMCLPRLPVGSLVSpecifications
AAH30958 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.74kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DSPG3/PGLB/Pg-Lb/SLRR3B | |
EPYC | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1833 | |
EPYC (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
APTLESINYDSETYDATLEDLDNLYNYENIPVGKVEIEIATVMPSGNRELLTPPPQPEKAQEEEEEEESTPRLIDGSSPQEPEFTGVLGPHTNEDFPTCLLCTCISTTVYCDDHELDAIPPLPKNTAYFYSRFNRIKKINKNDFASLSDLKRIDLTSNLISEIDEDAFRKLPQLRELVLRDNKIRQLPELPTTLTFIDISNNRLGRKGIKQEAFKDMYDLHHLYLTDNNLDHIPLPLPENLRALHLQNNNILEMHEDTFCNVKNLTYIRKALEDIRLDGNPINLSKTPQAYMCLPRLPVGSLV | |
RUO | |
EPYC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |