missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPB42 Partial ORF (NP_000110.1, 623 a.a. - 721 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002038-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127694

  • 4115.00 NOK / 10µg
Estimated Shipment: 19-09-2024
to see stock.

Description

Description

Erythrocyte membrane protein band 4.2 is an ATP-binding protein which may regulate the association of protein 3 with ankyrin. It probably has a role in erythrocyte shape and mechanical property regulation. Mutations in the EPB42 gene are associated with recessive spherocytic elliptocytosis and recessively transmitted hereditary hemolytic anemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSA
Specifications

Specifications

NP_000110.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KMPEKAEQYQPLTASVSLQNSLDAPMEDCVISILGRGLIHRERSYRFRSVWPENTMCAKFQFTPTHVGLQRLTVEVDCNMFQNLTNYKSVTVVAPELSA
RUO
EPB42
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2038
EPB42 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC116735/MGC116737/PA
EPB42
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers