Learn More
Abnova™ Human EOMES Partial ORF (NP_005433, 547 a.a. - 645 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008320-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar gene disrupted in mice is shown to be essential during trophoblast development and gastrulation. [provided by RefSeq]
Sequence: PYGIKSLPLQTSHALGYYPDPTFPAMAGWGGRGSYQRKMAAGLPWTSRTSPTVFSEDQLSKEKVKEEIGSSWIETPPSIKSLDSNDSGVYTSACKRRRLSpecifications
NP_005433 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PYGIKSLPLQTSHALGYYPDPTFPAMAGWGGRGSYQRKMAAGLPWTSRTSPTVFSEDQLSKEKVKEEIGSSWIETPPSIKSLDSNDSGVYTSACKRRRL | |
RUO | |
EOMES | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8320 | |
EOMES (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TBR2 | |
EOMES | |
Recombinant | |
wheat germ expression system |