Learn More
Abnova™ Human ENTPD2 Partial ORF (NP_982293.1, 187 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000954-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is the type 2 enzyme of the ecto-nucleoside triphosphate diphosphohydrolase family (E-NTPDase). E-NTPDases are a family of ecto-nucleosidases that hydrolyze 5'-triphosphates. This ecto-ATPase is an integral membrane protein. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]
Sequence: GRWFRPRKGTLGAMDLGGASTQITFETTSPAEDRASEVQLHLYGQHYRVYTHSFLCYGRDQVLQRLLASALQTHGFHPCWPRGFSTQVLSpecifications
NP_982293.1 | |
Liquid | |
954 | |
ENTPD2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD39L1/NTPDase-2 | |
ENTPD2 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GRWFRPRKGTLGAMDLGGASTQITFETTSPAEDRASEVQLHLYGQHYRVYTHSFLCYGRDQVLQRLLASALQTHGFHPCWPRGFSTQVL | |
RUO | |
ENTPD2 | |
Wheat Germ (in vitro) | |
GST |