missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ENO3 Full-length ORF (AAH17249, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002027-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134551

  • 4115.00 NOK / 10µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme, a homodimer, is found in skeletal muscle cells in the adult. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene can be associated with metabolic myopathies that may result from decreased stability of the enzyme. Two transcripts have been identified for this gene that differ only in their 5' UTR. [provided by RefSeq]

Sequence: MAMQKIFAREILDSRGNPTVEVDLHTAKGRFRAAVPSGASTGIYEALELRDGDKGRYLGKGVLKAVENINSTLGPALLQKKLSVADQEKVDKFMIELDGTENKSKFGANAILGVSLAVCKAGAAEKGVPLYRHIADLAGNPDLILPVPAFNVINGGSHAGNKLAMQEFMILPVGASSFKEAMRIGAEVYHHLKGVIKAKYGKDATNVGDEGGFAPNILENNEALELLKTAIQAAGYPDKVVIGMDVAASEFYRNGKYDLDFKSPDDPARHITGEKLGELYKSFIKNYPVVSIEDPFDQDDWATWTSFLSGVNIQIVGDDLTVTNPKRIAQAVEKKACNCLLLKVNQIGSVTESIQACKLAQSNGWGVMVSHRSGETEDTFIADLVVGLCTGQIKTGAPCRSERLAKYNQLMRIEEALGDKAIFAGRKFRNPKAK
Specifications

Specifications

AAH17249
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.48kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MSE
ENO3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2027
ENO3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMQKIFAREILDSRGNPTVEVDLHTAKGRFRAAVPSGASTGIYEALELRDGDKGRYLGKGVLKAVENINSTLGPALLQKKLSVADQEKVDKFMIELDGTENKSKFGANAILGVSLAVCKAGAAEKGVPLYRHIADLAGNPDLILPVPAFNVINGGSHAGNKLAMQEFMILPVGASSFKEAMRIGAEVYHHLKGVIKAKYGKDATNVGDEGGFAPNILENNEALELLKTAIQAAGYPDKVVIGMDVAASEFYRNGKYDLDFKSPDDPARHITGEKLGELYKSFIKNYPVVSIEDPFDQDDWATWTSFLSGVNIQIVGDDLTVTNPKRIAQAVEKKACNCLLLKVNQIGSVTESIQACKLAQSNGWGVMVSHRSGETEDTFIADLVVGLCTGQIKTGAPCRSERLAKYNQLMRIEEALGDKAIFAGRKFRNPKAK
RUO
ENO3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers